CAS RN 334981-10-1
IUPAC Navn:
4-[(pyrrolidine-1-sulfonyl)methyl]aniline
Synonymer:
4-[(pyrrolidine-1-sulfonyl)methyl]aniline
Molekylvægt (g/mol):
240.32
Molekylformel:
C11H16N2O2S
InChi Key:
VNSKHALYBQZMFW-UHFFFAOYSA-N
SMILES:
NC1=CC=C(CS(=O)(=O)N2CCCC2)C=C1
1
–
4
af
4
Resultater
1
1-((4-Aminobenzenemethane)sulfonyl)pyrrolidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-(Pyrrolidin-1-ylsulfonylmethyl)aniline, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
1-((4-Aminobenzenemethane)sulfonyl)pyrrolidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
1-((4-Aminobenzenemethane)sulfonyl)pyrrolidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.