CAS RN 331771-20-1
IUPAC Navn:
N-[4-({6-methoxy-7-[3-(morpholin-4-yl)propoxy]quinazolin-4-yl}amino)phenyl]benzamide
Synonymer:
N-[4-({6-methoxy-7-[3-(morpholin-4-yl)propoxy]quinazolin-4-yl}amino)phenyl]benzamide
Molekylvægt (g/mol):
513.6
Molekylformel:
C29H31N5O4
InChi Key:
OGNYUTNQZVRGMN-UHFFFAOYSA-N
SMILES:
COC1=C(OCCCN2CCOCC2)C=C2N=CN=C(NC3=CC=C(NC(=O)C4=CC=CC=C4)C=C3)C2=C1
1
–
3
af
3
Resultater
2
ZM 447439, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
ZM-447439, MedChemExpress
MedChemExpress ZM-447439 is an aurora kinase inhibitor with IC50s of 110 and 130 nM for aurora A and B, respectively.