CAS RN 2378-02-1
IUPAC Navn:
1,1,1,3,3,3-hexafluoro-2-(trifluoromethyl)propan-2-ol
Synonymer:
perfluoro-tert-butyl alcohol
Molekylvægt (g/mol):
236.04
Molekylformel:
C4HF9O
InChi Key:
XZNOAVNRSFURIR-UHFFFAOYSA-N
SMILES:
OC(C(F)(F)F)(C(F)(F)F)C(F)(F)F