Få mere at vide
Perfluoro-tert-butanol, 99%
Artikelnummer. 11490587
Skift visning
Mængde:
1 g
5 g
25 g
Pakningsstørrelse:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11490587 5 g 5g
11480587 1 g 1g
11400597 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 11490587 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. B22442.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
CAS: 2378-02-1 | C4HF9O | 236.04 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2378-02-1
MDL nummer MFCD00042092
Tekniske data
Kemisk navn eller materiale Perfluoro-tert-butanol
Smeltepunkt -17°C
Tæthed 1.693
Kogepunkt 44°C to 45°C
Flammepunkt None
Lugt Characteristic
Molekylær formel C4HF9O
Lineær formel (CF3)3COH
Brydningsindeks 1.3 00
Mængde 5 g
FN nummer UN2810
Beilstein 1841640
Synonym perfluoro-tert-butanol, nonafluoro-tert-butanol, perfluoro-tert-butyl alcohol, perfluoro-t-butanol, nonafluoro-tert-butyl alcohol, 2-propanol, 1,1,1,3,3,3-hexafluoro-2-trifluoromethyl, nonafluor-terc.butanol, 1,1,1,3,3,3-hexafluoro-2-trifluoromethyl propan-2-ol, 1,1,1,3,3,3-hexafluoro-2-trifluoromethyl-2-propanol, nonafluor-terc.butanol czech
InChI nøgle XZNOAVNRSFURIR-UHFFFAOYSA-N
SMIL C(C(F)(F)F)(C(F)(F)F)(C(F)(F)F)O
IUPAC navn 1,1,1,3,3,3-hexafluoro-2-(trifluoromethyl)propan-2-ol
Molekylvægt (g/mol) 236.04
PubChem CID 16924
Formel vægt 236.04
Procent renhed 99%