CAS RN 2135642-56-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
TAM-IN-2, MedChemExpress
MedChemExpress TAM-IN-2 is a TAM inhibitor extracted from patent US 20170275290 A1, pyrrolotriazine compound 0904.