CAS RN 2104-64-5
IUPAC Navn:
O-ethyl O-4-nitrophenyl phenylphosphonothioate
Synonymer:
santox EPN epn (insecticide)
Molekylvægt (g/mol):
323.3
Molekylformel:
C14H14NO4PS
InChi Key:
AIGRXSNSLVJMEA-UHFFFAOYNA-N
SMILES:
CCOP(=S)(OC1=CC=C(C=C1)[N+]([O-])=O)C1=CC=CC=C1
1
–
5
af
5
Resultater
1
EPN, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
EPN, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
EPN, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.