Få mere at vide
EPN, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | EPN |
| CAS | 2104-64-5 |
| Molekylær formel | C14 H14 N O4 P S |
| Mængde | 100 mg |
| Synonym | Phosphonothioic acid, phenyl-, O-ethyl O-(4-nitrophenyl) ester (9CI), Phosphonothioic acid, phenyl-, O-ethyl O-(p-nitrophenyl) ester (8CI), Phosphonothioic acid, phenyl-, ethyl p-nitrophenyl ester (7CI), EPN, EPN 300, NSC 404840, NSC 8943, O-ETHYL O-(P-NITROPHENYL) PHENYLPHOSPHONOTHIOATE, O-Ethyl O-(4-nitrophenyl) benzenethionophosphonate, O-Ethyl O-(4-nitrophenyl) phenylphosphonothioate, O-Ethyl O-(p-nitrophenyl) phenylphosphonothioate, O-Ethyl O-p-nitrophenyl benzenethiophosphonate, Santox |
| InChI formel | InChI=1S/C14H14NO4PS/c1-2-18-20(21,14-6-4-3-5-7-14)19-13-10-8-12(9-11-13)15(16)17/h3-11H,2H2,1H3 |
| SMIL | CCOP(=S)(Oc1ccc(cc1)[N+](=O)[O-])c2ccccc2 |
| IUPAC navn | ethoxy-(4-nitrophenoxy)-phenyl-sulfanylidene-λ^{5}-phosphane |
| Molekylvægt (g/mol) | 323.3 |
| Formel vægt | 323.0381 |