CAS RN 2055572-74-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(2R)-1-Azido-3-[[3-fluoro-4-(4-morpholinyl)phenyl]amino]-2-propanol 2-Acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(2R)-1-Azido-3-[[3-fluoro-4-(4-morpholinyl)phenyl]amino]-2-propanol 2-Acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(2R)-1-Azido-3-[[3-fluoro-4-(4-morpholinyl)phenyl]amino]-2-propanol 2-Acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.