CAS RN 173676-60-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(S)-5-Chloro-alpha-(2-cyclopropylethynyl)-2-[[(4-methoxyphenyl)methyl]amino]-alpha-(trifluoro-methyl)benzenemethanol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
(S)-5-Chloro-a-(cyclopropylethynyl)-2-[[(4-methoxyphenyl)methyl]amino]-a-(trifluoromethyl)benzenemethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(S)-5-Chloro-Alpha-(cyclopropylethynyl)-2-[[(4-methoxyphenyl)methyl]amino]-Alpha-(trifluoromethyl)benzenemethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.