CAS RN 172922-91-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Tocris Bioscience™ FICZ
High affinity aryl hydrocarbon receptor (AhR) agonist
2
FICZ, MedChemExpress
MedChemExpress FICZ is a potent aryl hydrocarbon receptor (AhR) agonist with a Kd of 70 pM.
3