Få mere at vide
Tocris Bioscience™ FICZ
High affinity aryl hydrocarbon receptor (AhR) agonist
Brand: Tocris Bioscience™ 5304/1
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
FICZ is a high affinity aryl hydrocarbon receptor (AhR) agonist (Kd = 70 pM). Proposed to be an endogenous AhR ligand. Induces transient expression of cytochrome P450-1A1 (CYP1A1) in vitro.Tekniske data
| FICZ | |
| 5,11-Dihydroindolo[3,2-b]carbazole-6-carboxaldehyde | |
| Soluble to 25mM in DMSO | |
| Bioactive Small Molecule |
| 172922-91-7 | |
| 1 mg | |
| Solid |
Produkttitel
Ser du en mulighed for forbedring?
Research Use Only. Not For Human or Veterinary Use