Få mere at vide
Tocris Bioscience™ FICZ
GREENER_CHOICE
High affinity aryl hydrocarbon receptor (AhR) agonist
Brand:  Tocris Bioscience™ 5304/1
Encompass
Produktkode 15766675
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
FICZ is a high affinity aryl hydrocarbon receptor (AhR) agonist (Kd = 70 pM). Proposed to be an endogenous AhR ligand. Induces transient expression of cytochrome P450-1A1 (CYP1A1) in vitro.
Tekniske data
Tekniske data
FICZ
5,11-Dihydroindolo[3,2-b]carbazole-6-carboxaldehyde
Soluble to 25mM in DMSO
Bioactive Small Molecule
172922-91-7
1 mg
Solid
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner
Produkttitel
Vælg et problem
Research Use Only. Not For Human or Veterinary Use