Få mere at vide
Tocris Bioscience™ FICZ
Beskrivelse
FICZ is a high affinity aryl hydrocarbon receptor (AhR) agonist (Kd = 70 pM). Proposed to be an endogenous AhR ligand. Induces transient expression of cytochrome P450-1A1 (CYP1A1) in vitro.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | FICZ |
| CAS | 172922-91-7 |
| Synonym | 5,11-Dihydroindolo[3,2-b]carbazole-6-carboxaldehyde |
| Mængde | 1 mg |
| Opløselighed | Soluble to 25mM in DMSO |
| Molekylær formel | C19H12N2O |
| Formel vægt | 284.31 |
| Fysisk form | Solid |
| Produkttype | Bioactive Small Molecule |
| Renhed | >95% |
Research Use Only. Not For Human or Veterinary Use