CAS RN 1672662-07-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
(Rac)-PT2399, MedChemExpress
MedChemExpress (Rac)-PT2399 (Compound 10e), the racemate of PT2399, acts as a potent and specific hypoxia-inducible factor 2a (HIF-2α) inhibitor with an IC50 of 0.01 μM.