CAS RN 1632-89-9
IUPAC Navn:
(1,2,2,2-²H₄)acetaldehyde
Synonymer:
(1,2,2,2-²H₄)acetaldehyde
Molekylvægt (g/mol):
48.08
Molekylformel:
C2H4O
InChi Key:
IKHGUXGNUITLKF-MZCSYVLQSA-N
SMILES:
[2H]C(=O)C([2H])([2H])[2H]
1
–
4
af
4
Resultater
1
Acetaldehyde-d4, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Acetaldehyde-d4, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Acetaldehyde-D4 - cargo aircraft only -, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Acetaldehyde-d4, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.