Få mere at vide
Acetaldehyde-D4 - cargo aircraft only -, TRC
Artikelnummer. 30323390
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30323390 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323390 Leverandør Toronto Research Chemicals Leverandørnr. TRCA132602250MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetaldehyde-d4
CAS 1632-89-9
Molekylær formel C2 2H4 O
Mængde 250 mg
Synonym Acetaldehyde-1,2,2,2-d4 (ACI), Acetaldehyde-d4 (6CI, 7CI, 8CI, 9CI), 1,2,2,2-Tetradeuterioethanone, Ethanal-d4, Perdeuterioacetaldehyde, Perdeuteroacetaldehyde, Tetradeuteroacetaldehyde, Acetaldehyde-D4
InChI formel InChI=1S/C2H4O/c1-2-3/h2H,1H3/i1D3,2D
SMIL [2H]C(=O)C([2H])([2H])[2H]
IUPAC navn 1,2,2,2-tetradeuterioethanone
Molekylvægt (g/mol) 48.08
Formel vægt 48.0513