Få mere at vide
Acetaldehyde-d4, CDN
Artikelnummer. 30315513
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30315513 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315513 Leverandør CDN Leverandørnr. CDND01631G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetaldehyde-d4
CAS 1632-89-9
Molekylær formel C2 2H4 O
Mængde 1 g
InChI formel InChI=1 S/C2H4O/c1-2-3/h2H,1H3/i1D3,2 D
SMIL [2 H]C(=O)C([2 H])([2 H])[2 H]
IUPAC navn 1,2,2,2-tetradeuterioethanone
Molekylvægt (g/mol) 48.08
Formel vægt 48.0513 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity