CAS RN 1628260-79-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
4
af
4
Resultater
1
GLPG1690, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
GLPG1690, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
GLPG1690, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Ziritaxestat, MedChemExpress
MedChemExpress Ziritaxestat (GLPG1690) is a first-in-class autotaxin (ATX) inhibitor, with an IC50 of 131 nM and a Ki of 15 nM.