Få mere at vide
Ziritaxestat, MedChemExpress
Artikelnummer. 16456287
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
25 mg
50 mg
100 mg
Pakningsstørrelse:
25mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16456287 5 mg -
16442776 25 mg 25mg
16439976 50 mg -
16434676 100 mg -
16413568 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 16456287 Leverandør Medchem Express Leverandørnr. HY101772
MedChemExpress Ziritaxestat (GLPG1690) is a first-in-class autotaxin (ATX) inhibitor, with an IC50 of 131 nM and a Ki of 15 nM.
Ziritaxestat (GLPG1690) is a first-in-class autotaxin (ATX) inhibitor, with an IC50 of 131 nM and a Ki of 15 nM.
Tekniske data
Kemisk navn eller materiale Ziritaxestat
CAS 1628260-79-6
Farve White
Renhedsgrad noter Research
Molekylær formel C30H33FN8O2S
Mængde 5 mg
Synonym GLPG1690
Opløselighedsinformation DMSO : 41.67 mg/mL (70.78 mM; Need ultrasonic)
SMIL N#CC1=C(C2=CC=C(F)C=C2)N=C(N(C3=C(CC)N=C4C(C)=CC(N5CCN(CC(N6CC(O)C6)=O)CC5)=CN43)C)S1
Molekylvægt (g/mol) 588.7
Formel vægt 588.7
Procent renhed 98.09%
Grad Research
Fysisk form Solid
Til brug med (applikation) COVID-19-immunoregulation