CAS RN 1600515-49-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
4
af
4
Resultater
1
Tocris Bioscience™ Tilfrinib
Potent and selective breast tumor kinase (Brk) inhibitor
2
Tilfrinib, MedChemExpress
MedChemExpress Tilfrinib (compound 4f) is a potent and selective inhibitor of breast tumor kinase (Brk) with an IC50 of 3.15 nM, which displays anti-proliferative activity and acts as a promising antitumor agent.
3
4