Få mere at vide
Tocris Bioscience™ Tilfrinib
GREENER_CHOICE
Potent and selective breast tumor kinase (Brk) inhibitor
Brand:  Tocris Bioscience™ 5579/10
Encompass
Produktkode 15796936
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
Tilfrinib is a potent and selective breast tumor kinase (Brk, also known as tyrosine-protein kinase 6, PTK6) inhibitor (IC50 = 3.15 nM). Exhibits >1000-fold selectivity for Brk over a panel of other kinases. Inhibits proliferation of breast cancer cells in vitro.
Tekniske data
Tekniske data
3-(9H-Pyrido[2,3-b]indol-4-ylamino)phenol
1600515-49-8
10 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner