Få mere at vide
Tocris Bioscience™ Tilfrinib
Beskrivelse
Tilfrinib is a potent and selective breast tumor kinase (Brk, also known as tyrosine-protein kinase 6, PTK6) inhibitor (IC50 = 3.15 nM). Exhibits >1000-fold selectivity for Brk over a panel of other kinases. Inhibits proliferation of breast cancer cells in vitro.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 3-(9H-Pyrido[2,3-b]indol-4-ylamino)phenol |
| CAS | 1600515-49-8 |
| Mængde | 50 mg |
| Mål | Other Kinase Inhibitors |
| Molekylær formel | C17H13N3O |
| Formel vægt | 275.3 |
| Renhed | >98% |