CAS RN 15965-31-8
IUPAC Navn:
5-chloro-1H-imidazole
Synonymer:
4-chloro-3H-imidazole
Molekylvægt (g/mol):
102.52
Molekylformel:
C3H3ClN2
InChi Key:
BQRBAXFOPZRMCU-UHFFFAOYSA-N
SMILES:
ClC1=CN=CN1
1
–
4
af
4
Resultater
2
4-Chloro-1H-imidazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-Chloro-1H-imidazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
4-Chloro-1H-imidazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.