CAS RN 1594092-37-1
IUPAC Navn:
N-[2-methoxy-4-(4-methylpiperazin-1-yl)phenyl]-4-{1-methyl-1H-pyrrolo[2,3-c]pyridin-3-yl}pyrimidin-2-amine
Synonymer:
N-[2-methoxy-4-(4-methylpiperazin-1-yl)phenyl]-4-{1-methylpyrrolo[2,3-c]pyridin-3-yl}pyrimidin-2-amine
Molekylvægt (g/mol):
429.53
Molekylformel:
C24H27N7O
InChi Key:
ZYVXTMKTGDARKR-UHFFFAOYSA-N
SMILES:
COC1=C(NC2=NC=CC(=N2)C2=CN(C)C3=CN=CC=C23)C=CC(=C1)N1CCN(C)CC1
1
–
4
af
4
Resultater
2
Tocris Bioscience™ AZ 191
Potent DYRK1B inhibitor
3
AZ191, MedChemExpress
MedChemExpress AZ191 is a potent inhibitor that selectively inhibits DYRK1B with IC50 of 17 nM.
4