CAS RN 1432660-47-3
IUPAC Navn:
3-[5-(cyclopropylsulfamoyl)-2-(thiophen-3-yl)phenyl]-1-[3-(trifluoromethyl)phenyl]urea
Synonymer:
3-[5-(cyclopropylsulfamoyl)-2-(thiophen-3-yl)phenyl]-1-[3-(trifluoromethyl)phenyl]urea
Molekylvægt (g/mol):
481.51
Molekylformel:
C21H18F3N3O3S2
InChi Key:
CCAWRGNYALGPQH-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC(NC(=O)NC2=C(C=CC(=C2)S(=O)(=O)NC2CC2)C2=CSC=C2)=CC=C1
1
–
2
af
2
Resultater
2
AGI-6780, MedChemExpress
MedChemExpress AGI-6780 that potently and selectively inhibits the tumor-associated mutant IDH2R140Q with IC50 of 23±1.7 nM. AGI-6780 is less potent against IDH2WT with IC50 of 190±8.1 nM.