CAS RN 1426138-42-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
FX1, MedChemExpress
MedChemExpress FX1 is a potent and specific BCL6 inhibitor, with an IC50 of around 35 μM.
2
Tocris Bioscience™ FX 1
Bcl-6 inhibitor; 10-fold more potent than endogenous Bcl-6 corepressors