CAS RN 141396-28-3
IUPAC Navn:
(2R,4R)-1-{5-[(diaminomethylidene)amino]-2-(3-methyl-1,2,3,4-tetrahydroquinoline-8-sulfonamido)pentanoyl}-4-methylpiperidine-2-carboxylic acid hydrate
Synonymer:
argatroban hydrate
Molekylvægt (g/mol):
526.65
Molekylformel:
C23H38N6O6S
InChi Key:
AIEZTKLTLCMZIA-LINCPPCXSA-N
SMILES:
O.C[C@@H]1CCN([C@H](C1)C(O)=O)C(=O)C(CCCN=C(N)N)NS(=O)(=O)C1=CC=CC2=C1NCC(C)C2
1
–
3
af
3
Resultater
1
Argatroban Monohydrate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Argatroban monohydrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Argatroban monohydrate, MedChemExpress
MedChemExpress Argatroban (monohydrate) (MD-805 (monohydrate)) is a direct, selective thrombin inhibitor.