CAS RN 1405-37-4
IUPAC Navn:
3,6-diamino-N-{[(8Z)-15-amino-11-(2-amino-3,4,5,6-tetrahydropyrimidin-4-yl)-8-[(carbamoylamino)methylidene]-2-(hydroxymethyl)-3,6,9,12,16-pentaoxo-1,4,7,10,13-pentaazacyclohexadecan-5-yl]methyl}hexanamide; sulfuric acid
Synonymer:
3,6-diamino-N-{[(8Z)-15-amino-11-(2-amino-3,4,5,6-tetrahydropyrimidin-4-yl)-8-[(carbamoylamino)methylidene]-2-(hydroxymethyl)-3,6,9,12,16-pentaoxo-1,4,7,10,13-pentaazacyclohexadecan-5-yl]methyl}hexanamide; sulfuric acid
Molekylvægt (g/mol):
766.79
Molekylformel:
C25H46N14O12S
InChi Key:
AJQVUIHGEOLMDY-SOCRLDLMNA-N
SMILES:
OS(O)(=O)=O.NCCCC(N)CC(=O)NCC1NC(=O)C(CO)NC(=O)C(N)CNC(=O)C(NC(=O)\C(NC1=O)=C\NC(N)=O)C1CCN=C(N)N1
1
–
5
af
5
Resultater
2
Capreomycin Sulfate, MP Biomedicals™
Often grouped with aminoglycosides. Capreomycin Sulfate, MP Biomedicals is a cyclic peptide antibiotic. This second line antibiotic binds across the ribosomal interface involving 23S rRNA helix 69 (H69) and 16S rRNA helix 44 (h44).
3
Capreomycin Sulfate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Capreomycin Sulfate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
5
Capreomycin sulfate, MedChemExpress
MedChemExpress Capreomycin sulfate is a peptide antibiotic, commonly grouped with the aminoglycosides, which is given in combination with other antibiotics for MDR-tuberculosis.