CAS RN 131631-89-5
IUPAC Navn:
N-(3-{4-[4-(2-oxo-1,2,3,4-tetrahydroquinolin-1-yl)piperidine-1-carbonyl]phenoxy}propyl)acetamide
Synonymer:
N-(3-{4-[4-(2-oxo-3,4-dihydroquinolin-1-yl)piperidine-1-carbonyl]phenoxy}propyl)acetamide
Molekylvægt (g/mol):
449.55
Molekylformel:
C26H31N3O4
InChi Key:
KSNUCNRMDYJBKT-UHFFFAOYSA-N
SMILES:
CC(=O)NCCCOC1=CC=C(C=C1)C(=O)N1CCC(CC1)N1C(=O)CCC2=CC=CC=C12
1
–
2
af
2
Resultater
1
Tocris Bioscience™ OPC 21268
CAS: 131631-89-5 Molekylær formel: C26H31N3O4
2
Fuscoside, MedChemExpress
MedChemExpress Fuscoside (OPC-21268) is an orally effective, nonpeptide, vasopressin V1 receptor antagonist with an IC50 of 0.4 μM.