Få mere at vide
Tocris Bioscience™ OPC 21268
Artikelnummer. 18074362
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18074362 10 mg
18015532 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18074362 Leverandør Tocris Bioscience™ Leverandørnr. 3924/10
Orally active vasopressin V1 antagonist
Nonpeptide vasopressin V1 receptor antagonist. Displays greater affinity for rat V1 than human V1 (Ki values are 25 and 8800 nM respectively). Exhibits 1000-fold selectivity for V1 receptors over V2 receptors. Orally active.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 131631-89-5
Molekylær formel C26H31N3O4
Tekniske data
Kemisk navn eller materiale OPC 21268
Mængde 10 mg
Formel vægt 449.54
Renhed >98%
Molekylvægt (g/mol) 449.551