CAS RN 1119449-37-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
4
af
4
Resultater
1
4-(3’,6’-Dimethyl-3’-heptyl)phenol Monoethoxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-(3’,6’-Dimethyl-3’-heptyl)phenol Monoethoxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-(3’,6’-Dimethyl-3’-heptyl)phenol Monoethoxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
4-(1-Ethyl-1,4-dimethylpentyl)-phenol-mono-ethoxylate, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality