Få mere at vide
4-(3’,6’-Dimethyl-3’-heptyl)phenol Monoethoxylate, TRC
Artikelnummer. 30374888
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30374888 50 mg
1 options
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Artikelnummer. 30374888 Leverandør Toronto Research Chemicals Leverandørnr. TRCD47311550MG
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(3’,6’-Dimethyl-3’-heptyl)phenol Monoethoxylate
CAS 1119449-37-4
Molekylær formel C17 H28 O2
Mængde 50 mg
Synonym 4-(1-Ethyl-1,4-dimethylpentyl)-phenol-mono-ethoxylate, Ethanol, 2-[4-(1-ethyl-1,4-dimethylpentyl)phenoxy]-, 2-[4-(1-Ethyl-1,4-dimethylpentyl)phenoxy]ethanol
InChI formel InChI=1S/C17H28O2/c1-5-17(4,11-10-14(2)3)15-6-8-16(9-7-15)19-13-12-18/h6-9,14,18H,5,10-13H2,1-4H3
SMIL CCC(C)(CCC(C)C)c1ccc(OCCO)cc1
IUPAC navn 2-[4-(3,6-dimethylheptan-3-yl)phenoxy]ethanol
Molekylvægt (g/mol) 264.4
Formel vægt 264.2089