Learn More
ABEBA™ White Occupational Shoes
Anti-static occupational surgical clogs are non-slip.
417.00 DKK - 485.00 DKK
Product Code | Brand | dimSize | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | dimSize | Price | Quantity & Availability | ||||||
15233715
|
ABEBA™
9600-37/38 |
468.00 DKK
1 pair |
Estimated Shipment: 08-07-2025
Log in to see stock. |
|||||||
15263715
|
ABEBA™
9600-43/44 |
459.00 DKK
1 pair |
Estimated Shipment: 20-08-2025
Log in to see stock. |
|||||||
15253715
|
ABEBA™
9600-41/42 |
485.00 DKK
1 pair |
Estimated Shipment: 24-07-2025
Log in to see stock. |
|||||||
15223715
|
ABEBA™
9600-35/36 |
468.00 DKK
1 pair |
Estimated Shipment: 08-07-2025
Log in to see stock. |
|||||||
15243715
|
ABEBA™
9600-39/40 |
456.00 DKK
1 pair |
Estimated Shipment: 03-07-2025
Log in to see stock. |
|||||||
15300202
|
ABEBA™
9600-45/46 |
417.00 DKK
1 pair |
Estimated Shipment: 05-08-2025
Log in to see stock. |
|||||||
Description
- Autoclavable at 121° C for 15 minutes (with no additives)
- White
- Antistatic
- Washable at 60° C
- Foldable, adjustable heel strap
- SRC slip resistance according to EN ISO 20347: 2012
- CE EN ISO 20347: 2012
Specifications
White | |
9600 | |
Controlled Environments Shoes |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
ABEBA™ White Occupational Shoes