Få mere at vide
3M™ White/Blue Combined SMS Laminate Protective Coveralls
Protective coveralls are manufactured from a high quality laminated material with a breathable back panel to improve air ventilation and reduce heat build-up.
Brand: 3M™ 7000089639
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
- Breathable back panel to help improve air ventilation and reduce heat build-up.
- Knit cuffs provide a soft wrist closure for increased comfort compared to elastic closures.
- Elastic waist and ankles for convenience and freedom of movement.
- Two-way zipper with sealable storm flap for convenience and to help provide additional protection.
Tekniske data
General Purpose Coveralls | |
Unisex | |
Elastic | |
Hooded | |
Category III | |
164 to 170 cm |
Small | |
Single-use | |
Knit | |
Non-sterile | |
4535W |
Korrektion af produktindhold
Produkttitel
3M™ White/Blue Combined SMS Laminate Protective Coveralls > Size: Small
Ser du en mulighed for forbedring?Del en indholdskorrektion