Learn More
Cytiva Whatman™ Mini-UniPrep™ Multicompressor and Accessories
Streamlines filtration, saves time, and reduces hand stress by enabling the filtration of eight Mini-UniPrep™ Vials at once. Trays to accomodate additional vials are sold separately.
175.00 DKK - 2805.00 DKK
Specifications
For Use With (Application) | HPLC/UHPLC |
---|---|
Product Line | Whatman™ |
Quantity | 1 Ea. |
Description
- Streamlines syringeless filtration by filtering up to eight filter vials simultaneously
- Designed for Cytiva's Mini-UniPrep™ Standard Plastic Filter Vial
- Mini-UniPrep™ Vials can be loaded directly into the autosampler
- The multiprocessor includes one blue 8-position tray
- Additional trays provide the added convenience of pre-loading up to eight Mini-UniPrep™ Filter Vials
Specifications
HPLC/UHPLC | |
Whatman™ | |
1 Ea. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Cytiva Whatman™ Mini-UniPrep™ Multicompressor and Accessories