missing translation for 'onlineSavingsMsg'
Få mere at vide

Tolmetin sodium salt dihydrate, 98+%, Thermo Scientific™

An NSAID compound

433.59 DKK - 4761.27 DKK

Kemiske identifikatorer

CAS 64490-92-2
Molekylær formel C15H18NNaO5
Molekylvægt (g/mol) 315.301
MDL nummer MFCD00150761
InChI nøgle QQILXENAYPUNEA-UHFFFAOYSA-M
Synonym tolmetin sodium dihydrate, sodium tolmetin dihydrate, tolumetin sodium dihydrate, unii-02n1tzf99f, sodium 2-1-methyl-5-4-methylbenzoyl-1h-pyrrol-2-yl acetate dihydrate, tolmetin sodium salt dihydrate, sodium 1-methyl-5-p-toluoylpyrrole-2-acetate dihydrate, tolectin tn, 1-methyl-5-p-toluoyl pyrrole-2 acetic acid
PubChem CID 23677829
ChEBI CHEBI:72014
IUPAC navn sodium;2-[1-methyl-5-(4-methylbenzoyl)pyrrol-2-yl]acetate;dihydrate
SMIL CC1=CC=C(C=C1)C(=O)C2=CC=C(N2C)CC(=O)[O-].O.O.[Na+]
Se flere specifikationer

Produkter 3
Produktkode Brand Mængde Pris Antal & tilgængelighed  
Produktkode Brand Mængde Pris Antal & tilgængelighed  
15474989
Dokumentvisning
Thermo Scientific Alfa Aesar
J62663.03
1g
433.59 DKK
1g
Ver productos alternativos
Ahorre hasta 
»
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
15484989
Ver documentos
Thermo Scientific Alfa Aesar
J62663.06
5g
1373.15 DKK
5g
Ver productos alternativos
Ahorre hasta 
»
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
15494989
Ver documentos
Thermo Scientific Alfa Aesar
J62663.14
25g
4761.27 DKK
25g
Ver productos alternativos
Ahorre hasta 
»
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Descripción

Descripción

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Tolmetin Sodium is an anti-inflammatory, antirheumatic agent. It can inhibit aldo-keto reductases (e.g., IC50 = 2.39 µM for human aldose reductase), a family of enzymes that catalyze the NADPH-dependent reduction of carbonyl groups in a number of important steroid, metabolic, and prostanoid molecules. It is a beta-adrenergic blocker.

Solubility
Soluble in water

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Especificaciones

Identificadores de productos químicos

64490-92-2
315.301
QQILXENAYPUNEA-UHFFFAOYSA-M
23677829
sodium;2-[1-methyl-5-(4-methylbenzoyl)pyrrol-2-yl]acetate;dihydrate
C15H18NNaO5
MFCD00150761
tolmetin sodium dihydrate, sodium tolmetin dihydrate, tolumetin sodium dihydrate, unii-02n1tzf99f, sodium 2-1-methyl-5-4-methylbenzoyl-1h-pyrrol-2-yl acetate dihydrate, tolmetin sodium salt dihydrate, sodium 1-methyl-5-p-toluoylpyrrole-2-acetate dihydrate, tolectin tn, 1-methyl-5-p-toluoyl pyrrole-2 acetic acid
CHEBI:72014
CC1=CC=C(C=C1)C(=O)C2=CC=C(N2C)CC(=O)[O-].O.O.[Na+]

Especificaciones

Tolmetin sodium salt dihydrate
C15H18NNaO5
14,9518
Soluble in water
CC1=CC=C(C=C1)C(=O)C2=CC=C(N2C)CC(=O)[O-].O.O.[Na+]
315.301
CHEBI:72014
≥98%
64490-92-2
MFCD00150761
tolmetin sodium dihydrate, sodium tolmetin dihydrate, tolumetin sodium dihydrate, unii-02n1tzf99f, sodium 2-1-methyl-5-4-methylbenzoyl-1h-pyrrol-2-yl acetate dihydrate, tolmetin sodium salt dihydrate, sodium 1-methyl-5-p-toluoylpyrrole-2-acetate dihydrate, tolectin tn, 1-methyl-5-p-toluoyl pyrrole-2 acetic acid
QQILXENAYPUNEA-UHFFFAOYSA-M
sodium;2-[1-methyl-5-(4-methylbenzoyl)pyrrol-2-yl]acetate;dihydrate
23677829
315.30 (279.27 Anhydrous)
Videos
Seguridad y manipulación

Seguridad y manipulación

GHS H Statement
H302-H315
Harmful if swallowed.
Causes skin irritation.

P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c

H302-H315-H319

missing translation for 'einecsNumber' : 252-687-3

missing translation for 'rtecsNumber' : UX9280000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documentos

Documentos

Product Certifications

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.