missing translation for 'onlineSavingsMsg'
Få mere at vide

Tolmetin sodium salt dihydrate, 98+%, Thermo Scientific™

An NSAID compound

Brand:  Thermo Scientific Alfa Aesar J62663.03

Mængde 1g

 Se flere versioner af dette produkt

Produktkode 15474989

  • 433.59 DKK / 1g

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolitik

Beskrivelse

Beskrivelse

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Tolmetin Sodium is an anti-inflammatory, antirheumatic agent. It can inhibit aldo-keto reductases (e.g., IC50 = 2.39 µM for human aldose reductase), a family of enzymes that catalyze the NADPH-dependent reduction of carbonyl groups in a number of important steroid, metabolic, and prostanoid molecules. It is a beta-adrenergic blocker.

Solubility
Soluble in water

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Tekniske data

Kemiske identifikatorer

64490-92-2
315.301
QQILXENAYPUNEA-UHFFFAOYSA-M
23677829
sodium;2-[1-methyl-5-(4-methylbenzoyl)pyrrol-2-yl]acetate;dihydrate
C15H18NNaO5
MFCD00150761
tolmetin sodium dihydrate, sodium tolmetin dihydrate, tolumetin sodium dihydrate, unii-02n1tzf99f, sodium 2-1-methyl-5-4-methylbenzoyl-1h-pyrrol-2-yl acetate dihydrate, tolmetin sodium salt dihydrate, sodium 1-methyl-5-p-toluoylpyrrole-2-acetate dihydrate, tolectin tn, 1-methyl-5-p-toluoyl pyrrole-2 acetic acid
CHEBI:72014
CC1=CC=C(C=C1)C(=O)C2=CC=C(N2C)CC(=O)[O-].O.O.[Na+]

Tekniske data

Tolmetin sodium salt dihydrate
C15H18NNaO5
1g
tolmetin sodium dihydrate, sodium tolmetin dihydrate, tolumetin sodium dihydrate, unii-02n1tzf99f, sodium 2-1-methyl-5-4-methylbenzoyl-1h-pyrrol-2-yl acetate dihydrate, tolmetin sodium salt dihydrate, sodium 1-methyl-5-p-toluoylpyrrole-2-acetate dihydrate, tolectin tn, 1-methyl-5-p-toluoyl pyrrole-2 acetic acid
QQILXENAYPUNEA-UHFFFAOYSA-M
sodium;2-[1-methyl-5-(4-methylbenzoyl)pyrrol-2-yl]acetate;dihydrate
23677829
315.30 (279.27 Anhydrous)
64490-92-2
MFCD00150761
14,9518
Soluble in water
CC1=CC=C(C=C1)C(=O)C2=CC=C(N2C)CC(=O)[O-].O.O.[Na+]
315.301
CHEBI:72014
≥98%
Produktforslag

Produktforslag

Videoer
SDS
Dokumenter

Dokumenter

Certifikater
Kampagner

Kampagner

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.