missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TEK (TIE2) [Y897S], GST Tag Recombinant Protein
Recombinant Protein
4245.00 DKK - 61400.00 DKK
Specifications
Accession Number | Q02763 |
---|---|
Concentration | See Label |
For Use With (Application) | Kinase Assay |
Formulation | 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5 |
Gene ID (Entrez) | 7010 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15920293
|
Invitrogen™
A30519 |
10 ÎĽg |
4245.00 DKK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17118745
|
Invitrogen™
A32878 |
100 ÎĽg |
19780.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17148785
|
Invitrogen™
A33368 |
1 mg |
61400.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TEK (TIE2) Y897S receptor tyrosine kinase is expressed principally on vascular endothelium. Disrupting TEK Y897S function in mice results in embryonic lethality with defects in embryonic vasculature, suggesting a role in blood vessel maturation and maintenance. Angiopoietin-1 is a secreted growth factor that binds to and activates the TEK receptor tyrosine kinase. SHP2 and GRB2 are recruited to the activated TEK kinase domain and are part of the cellular responses that mediate TEK function. TEK expression is upregulated in the endothelium of vascular 'hot spots' in human breast cancer specimens. However, TEK is also overexpressed in areas of active angiogenesis in normal tissues.Specifications
Q02763 | |
Kinase Assay | |
7010 | |
Human TEK (TIE2) [Y897S], GST Tag | |
TIE2 (TEK) (Y897S Mutant) | |
771-end | |
Human | |
GST-tag | |
Dry Ice | |
Liquid |
See Label | |
50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5 | |
66.7 kDa | |
TIE2 | |
TIE Kinase Family | |
Unconjugated | |
Recombinant | |
-80°C, Avoid Freeze/Thaw Cycles | |
Insect cells |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Human TEK (TIE2) [Y897S], GST Tag Recombinant Protein