missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Human TEK (TIE2) [Y897S], GST Tag Recombinant Protein
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 μg
100 μg
Pakningsstørrelse:
10µg
Beskrivelse
TEK (TIE2) Y897S receptor tyrosine kinase is expressed principally on vascular endothelium. Disrupting TEK Y897S function in mice results in embryonic lethality with defects in embryonic vasculature, suggesting a role in blood vessel maturation and maintenance. Angiopoietin-1 is a secreted growth factor that binds to and activates the TEK receptor tyrosine kinase. SHP2 and GRB2 are recruited to the activated TEK kinase domain and are part of the cellular responses that mediate TEK function. TEK expression is upregulated in the endothelium of vascular 'hot spots' in human breast cancer specimens. However, TEK is also overexpressed in areas of active angiogenesis in normal tissues.

Tekniske data
Tekniske data
Adgangsnummer | Q02763 |
Koncentration | See Label |
Til brug med (applikation) | Kinase Assay |
Formulering | 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5 |
Gen-id (Entrez) | 7010 |
Molekylvægt (g/mol) | 66.7 kDa |
Navn | Human TEK (TIE2) [Y897S], GST Tag |
Mængde | 1 mg |
Gene Alias | TIE2 |
Fælles navn | TIE2 (TEK) (Y897S Mutant) |
Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Invitrogen™ Human TEK (TIE2) [Y897S], GST Tag Recombinant Protein >
Ser du en mulighed for forbedring?Del en indholdskorrektion