Learn More
Interscience™ Support for Eppendorf™ pipettors
Support stand for Eppendorf™ pipettors.
759.00 DKK
Specifications
Type Support Stand
For Use With (Application) Holding Pipettor
View More Specs
Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
17272254
View Documents
Interscience™
413016
easySpiral, easySpiral Pro (5 mL)
759.00 DKK
Each
View alternative products
Save Up to 
»
17806706
View Documents
Interscience™
413017
easySpiral, easySpiral Pro (1.5 mL and 2 mL)
759.00 DKK
Each
View alternative products
Save Up to 
»
Description
Description
  • Supports pipettors of various sizes
Specifications
Specifications
Support Stand
Holding Pipettor
Documents
Product Certifications
Product Content Correction
Product Title
Interscience™ Support for Eppendorf™ pipettors