Learn More
Ohaus™ Stainless Steel Calibration Weights - OIML E1
Adjusted to Meet or Exceed OIML E1 Tolerances
1250.00 DKK - 33580.00 DKK
Specifications
Certifications/Compliance | No Certificate |
---|---|
Material | Troemner Alloy 8 |
Includes | Protective Case |
Product Code | Brand | Mass | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | ||||||
16642312
|
Ohaus™
30517972 |
1 g |
1250.00 DKK
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
16652312
|
Ohaus™
30517973 |
2 g |
1280.00 DKK
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
16662312
|
Ohaus™
30517974 |
5 g |
1310.00 DKK
Each |
In Stock
Log in to see stock. |
||||||
16672312
|
Ohaus™
30517975 |
10 g |
1435.00 DKK
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
16682312
|
Ohaus™
30517976 |
20 g |
1510.00 DKK
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
16692312
|
Ohaus™
30517977 |
50 g |
1800.00 DKK
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
16602322
|
Ohaus™
30517978 |
100 g |
2130.00 DKK
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
16612322
|
Ohaus™
30517979 |
200 mg |
2515.00 DKK
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
16622322
|
Ohaus™
30517980 |
500 g |
3550.00 DKK
Each |
In Stock
Log in to see stock. |
||||||
16632322
|
Ohaus™
30517981 |
1 kg |
5435.00 DKK
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
16642322
|
Ohaus™
30517982 |
2 kg |
8000.00 DKK
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
16652322
|
Ohaus™
30517983 |
5 kg |
13355.00 DKK
Each |
In Stock
Log in to see stock. |
||||||
16662322
|
Ohaus™
30517984 |
10 kg |
19090.00 DKK
Each |
In Stock
Log in to see stock. |
||||||
16672322
|
Ohaus™
30517985 |
20 kg |
33580.00 DKK
Each |
In Stock
Log in to see stock. |
||||||
Description
- OIML Class E1 offers high accuracy with tight tolerances and guaranteed uncertainty 1/3 of the tolerance
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish
- Weights ranging in masses from 1 gram to 20 kilograms are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free, protecting your investment
Specifications
No Certificate | |
Troemner Alloy 8 | |
Protective Case |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ohaus™ Stainless Steel Calibration Weights - OIML E1