missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress SPQ

MedChemExpress SPQ is being used to examine and measure membrane chloride transport mechanisms.

Encompass_Preferred

523.71 DKK - 835.91 DKK

Specifications

Storage Requirements 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Solubility H2O : 25 mg/mL (88.86 mM; Need ultrasonic) ∣ DMSO : < 1 mg/mL (insoluble or slightly soluble)
Shipping Condition Room temperature
CAS 83907-40-8
Molecular Weight (g/mol) 281.33 g/mol
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16464687
Encompass_Preferred
View Documents
MedChemExpress
HY-D0936
50 mg
523.71 DKK
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16438916
Encompass_Preferred
View Documents
MedChemExpress
HY-D0936
100 mg
835.91 DKK
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Room temperature
281.33 g/mol
H2O : 25 mg/mL (88.86 mM; Need ultrasonic) ∣ DMSO : < 1 mg/mL (insoluble or slightly soluble)
83907-40-8
HBV
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
MedChemExpress SPQ

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.