Learn More
Seratrodast, MedChemExpress
MedChemExpress Seratrodast(AA 2414) is an anti-asthmatic agent and a potent and selective thromboxane A2 receptor (TP) antagonist.

443.14 DKK - 1117.91 DKK
Specifications
Chemical Name or Material | Seratrodast |
---|---|
Physical Form | Solid |
CAS | 112665-43-7 |
Color | Orange |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16484477
![]() |
Medchem Express
HY-B0774 |
50 mg |
1117.91 DKK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16428608
![]() |
Medchem Express
HY-B0774 |
10 mg |
443.14 DKK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Seratrodast(AA 2414) is an anti-asthmatic agent and a potent and selective thromboxane A2 receptor (TP) antagonist.Specifications
Seratrodast | |
112665-43-7 | |
Research | |
COVID-19-immunoregulation | |
DMSO : ≥ 100 mg/mL (282.14 mM) | |
354.44 | |
99.68% |
Solid | |
Orange | |
C22H26O4 | |
AA 2414 | |
CC(C1=O)=C(C(C(C)=C1C)=O)C(C2=CC=CC=C2)CCCCCC(O)=O | |
354.44 | |
Research |
Safety and Handling
H361
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.