missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Selpercatinib

Potent and selective RET inhibitor
1455.00 DKK - 5425.00 DKK
Specifications
Chemical Name or Material | 6-(2-Hydroxy-2-methylpropoxy)-4-[6-[6-[(6-methoxy-3-pyridinyl)methyl]-3,6-diazabicyclo[3.1.1 ]hept-3-yl]-3-pyridinyl]pyrazolo[1,5-a]pyridine-3-carbonitrile |
---|---|
CAS | 2152628-33-4 |
Target | Ret |
Molecular Formula | C29H31N7O3 |
Purity | 0.98 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18747026
![]() |
Tocris Bioscience™
7696/10 |
10 mg |
1455.00 DKK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18759196
![]() |
Tocris Bioscience™
7696/50 |
50 mg |
5425.00 DKK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Selpercatinib, is a potent and selective receptor tyrosine kinase RET inhibitor (EC50 = 4 nM). It shows > 100-fold selectivity for > 95% of 228 purified kinase domains in radiometric assays. In vitro, it inhibits proliferation of cancer cell lines containing RET mutations or gene fusions. In vivo, it inhibits tumor growth in diverse RET mouse models. Selpercatinib is orally bioavailable.Specifications
6-(2-Hydroxy-2-methylpropoxy)-4-[6-[6-[(6-methoxy-3-pyridinyl)methyl]-3,6-diazabicyclo[3.1.1 ]hept-3-yl]-3-pyridinyl]pyrazolo[1,5-a]pyridine-3-carbonitrile | |
Ret | |
0.98 |
2152628-33-4 | |
C29H31N7O3 |
Safety and Handling
Recommended Storage : Store at -20°C
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tocris Bioscience™ Selpercatinib