Learn More
uvex Rubiflex S NB27B Chemical Protection Gloves
Very lightweight, stockinette NBR chemical protection gloves provide protection from a wide range of chemical and mechanical risks. Gloves offer a high level of tactile sensitivity to make them compatible with touch displays.
97.20 DKK - 980.00 DKK
Specifications
Color | Blue |
---|---|
Latex Free | Yes |
Material | Nitrile |
Product Type | Chemical-resistant Gloves |
EN388 Standard | 2111X |
Description
- Very lightweight, stockinette NBR chemical protection gloves
- Fully coated with NBR coating, approximately 0.40 mm thick
- High quality interlock cotton supporting material increases user comfort
- Resistant to grease, mineral oils, and a wide variety of chemicals
- Enhanced grip in damp or wet areas
- Suitable for operating touch displays and food processing
- Free of harmful substances in accordance with OEKO-TEX™ Standard 100
Specifications
Blue | |
Nitrile | |
2111X | |
JKNOPT | |
Gauntlet |
Yes | |
Chemical-resistant Gloves | |
Type A | |
Automotive, Chemical Handling, Food Processing, Laboratory Work, and Painting | |
Interlock Cotton |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
uvex Rubiflex S NB27B Chemical Protection Gloves