Learn More
uvex Rubiflex ESD NB35A Chemical Protection Gloves
Lightweight, anti-static stockinette NBR chemical protection gloves are ideal for applications when high electrostatic discharge properties are required, in addition to protection from grease, mineral oils, and various chemicals.
1455.00 DKK - 1520.00 DKK
Specifications
Certifications/Compliance (Europe) DIN EN 16350:2014
Applications Laboratory
Coating Material Nitrile
Color Black
Disposable Reusable
View More Specs
Products 6
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
17720988
View Documents
uvex
6095406
6
1520.00 DKK
10 pairs
Only null left
Estimated Shipment: 17-07-2025
Add to basket
17200511
View Documents
uvex
6095407
7
1455.00 DKK
10 pairs
Only null left
Estimated Shipment: 17-07-2025
Add to basket
17210511
View Documents
uvex
6095408
8
1455.00 DKK
10 pairs
Only null left
Estimated Shipment: 17-07-2025
Add to basket
17220511
View Documents
uvex
6095409
9
1455.00 DKK
10 pairs
Only null left
Estimated Shipment: 17-07-2025
Add to basket
17230511
View Documents
uvex
6095410
10
1455.00 DKK
10 pairs
Only null left
Estimated Shipment: 17-07-2025
Add to basket
17240511
View Documents
uvex
6095411
11
1455.00 DKK
10 pairs
Only null left
Estimated Shipment: 17-07-2025
Add to basket
Description
Description
  • Anti-static stockinette chemical protection gloves
  • Fully coated with special conductive, 0.40 mm NBR coating
  • Cotton interlock/carbon supporting material increases comfort
  • Ergonomic fit is extremely flexible for tactile sensitivity
  • Meets the current standard DIN EN 16350:2014
  • Contact resistance less than 108 ohms
  • Enhanced grip in damp or wet areas
  • Resistant to grease, mineral oils, and various chemicals
  • Free of harmful substances in accordance with OEKO-TEX™ Standard 100
  • Ideal for use in potentially explosive atmospheres and ESD environments
Specifications
Specifications
DIN EN 16350:2014
Nitrile
Reusable
Ambidextrous
NBR
ESD Gloves
2111X
JKLOPT
Unisex
Cotton, Carbon
35 cm
Laboratory
Black
Smooth
Yes
Powder Free
Non-sterile
Type A
Automotive, Chemical Handling, Electrostatic Discharge, Plastic Processing, and Refineries
Gauntlet
Rubiflex ESD NB35A
Documents
Product Certifications
Product Content Correction
Product Title
uvex Rubiflex ESD NB35A Chemical Protection Gloves