Learn More
enQuireBio™ Recombinant Mouse Major urinary protein 11 Protein
A DNA sequence encoding the Mus musculus (Mouse) Major urinary protein 11, was expressed in the hosts and tags indicated.
1820.00 DKK - 22245.00 DKK
Specifications
Accession Number | NP_001157998.1 |
---|---|
Gene ID (Entrez) | 100039028 |
Name | Major urinary protein 11 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16068440
|
enQuireBio™
QP7360-YE-100UG |
100 μg |
5015.00 DKK
100µg |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16088440
|
enQuireBio™
QP7360-YE-1MG |
1 mg |
22245.00 DKK
1mg |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16078440
|
enQuireBio™
QP7360-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098440
|
enQuireBio™
QP7360-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16008450
|
enQuireBio™
QP7360-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16018450
|
enQuireBio™
QP7360-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001157998.1 | |
Major urinary protein 11 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
100039028 | |
Research Use Only | |
Mup11 | |
Recombinant Protein | |
Yeast | |
REKINGEWHT IILASDKREK IEDNGNFRLF LEQIHVLENS LVLKFHTVRD EECSELSMVA DKTEKAGEYS VTYDGFNTFT IPKTDYDNFL MAHLINEKDG ETFQLMGLYG REPDLSSDIK ERFAQLCEEH GILRENIIDL SNANRCLQAR E | |
Greater than 81.5% as determined by SDS-PAGE. |