Få mere at vide
enQuireBio™ Recombinant Mouse Major urinary protein 11 Protein
Artikelnummer. 16008450
Mængde:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Pakningsstørrelse:
100µg
10µg
1mg
200µg
500µg
50µg
Artikelnummer. 16008450
Brand: enQuireBio™ QP7360ye500ug
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
A DNA sequence encoding the Mus musculus (Mouse) Major urinary protein 11, was expressed in the hosts and tags indicated.
Tekniske data
Adgangsnummer NP_001157998.1
Gen-id (Entrez) 100039028
Navn Major urinary protein 11 Protein
Mængde 500 μg
Regulatorisk status Research Use Only
Endotoksin koncentration Not determined.
Gen symbol Mup11
Protein længde Full Length
Produkttype Recombinant Protein
Krydsreaktivitet Mouse
Arter Yeast
Protein tag His
Sekvens REKINGEWHT IILASDKREK IEDNGNFRLF LEQIHVLENS LVLKFHTVRD EECSELSMVA DKTEKAGEYS VTYDGFNTFT IPKTDYDNFL MAHLINEKDG ETFQLMGLYG REPDLSSDIK ERFAQLCEEH GILRENIIDL SNANRCLQAR E
Buffer Tris-based buffer, 50% glycerol
Renhed eller kvalitetsklasse Greater than 81.5% as determined by SDS-PAGE.