missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Hemoglobin subunit beta-2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Hemoglobin subunit beta-2, was expressed in the hosts and tags indicated.
1820.00 DKK - 24645.00 DKK
Specifications
Accession Number | NP_058652.1 |
---|---|
Gene ID (Entrez) | 15130 |
Name | Hemoglobin subunit beta-2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16078350
|
enQuireBio™
QP7346-ec-10ug |
10 ÎĽg | N/A | ||||||
16018360
|
enQuireBio™
QP7346-ec-50ug |
50 ÎĽg | N/A | ||||||
16068350
|
enQuireBio™
QP7346-ec-100ug |
100 ÎĽg |
5555.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16098350
|
enQuireBio™
QP7346-ec-200ug |
200 ÎĽg | N/A | ||||||
16008360
|
enQuireBio™
QP7346-ec-500ug |
500 ÎĽg | N/A | ||||||
16088350
|
enQuireBio™
QP7346-ec-1mg |
1 mg |
24645.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_058652.1 | |
Hemoglobin subunit beta-2 Protein | |
Not determined. | |
Full Length | |
Mouse | |
GST | |
Tris-based buffer, 50% glycerol |
15130 | |
Research Use Only | |
Hbb-b2 | |
Recombinant Protein | |
E. coli | |
VHLTDAEKSA VSCLWAKVNP DEVGGEALGR LLVVYPWTQR YFDSFGDLSS ASAIMGNPKV KAHGKKVITA FNEGLKNLDN LKGTFASLSE LHCDKLHVDP ENFRLLGNAI VIVLGHHLGK DFTPAAQAAF QKVVAGVATA LAHKYH | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse Hemoglobin subunit beta-2 Protein