missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Hemoglobin subunit beta-2 Protein

A DNA sequence encoding the Mus musculus (Mouse) Hemoglobin subunit beta-2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7346-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16008360

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_058652.1
Hemoglobin subunit beta-2 Protein
Research Use Only
Hbb-b2
Recombinant Protein
E. coli
VHLTDAEKSA VSCLWAKVNP DEVGGEALGR LLVVYPWTQR YFDSFGDLSS ASAIMGNPKV KAHGKKVITA FNEGLKNLDN LKGTFASLSE LHCDKLHVDP ENFRLLGNAI VIVLGHHLGK DFTPAAQAAF QKVVAGVATA LAHKYH
Greater than 90% as determined by SDS-PAGE.
15130
500 μg
Not determined.
Full Length
Mouse
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers