missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Hemoglobin subunit beta-2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Hemoglobin subunit beta-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7346-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_058652.1 | |
Hemoglobin subunit beta-2 Protein | |
Research Use Only | |
Hbb-b2 | |
Recombinant Protein | |
E. coli | |
VHLTDAEKSA VSCLWAKVNP DEVGGEALGR LLVVYPWTQR YFDSFGDLSS ASAIMGNPKV KAHGKKVITA FNEGLKNLDN LKGTFASLSE LHCDKLHVDP ENFRLLGNAI VIVLGHHLGK DFTPAAQAAF QKVVAGVATA LAHKYH | |
Greater than 90% as determined by SDS-PAGE. |
15130 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
GST | |
Tris-based buffer, 50% glycerol |